Manganese(II) nitrate tetrahydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Manganese dinitrate tetrahydrate
, Manganous nitrate tetrahydrate
222130-1KG
222130-250G
BT222130-1KGEA
179.83
USD
BT222130-1KG
BT222130-250G
Manganese(II) nitrate tetrahydrate ≥98%
Manganese (II) nitrate tetrahydrate
- Hygroscopic
- Crystalline
Formula:
Mn(NO₃)₂·4H₂O MW: 251.01 g/mol Melting Pt: 26 °C |
MDL Number:
MFCD00149789 CAS Number: 20694-39-7 UN: 2724 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...