2-Aminoacetonitrile hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Aminoacetonitrile hydrochloride

222045-25G
BT222045-25GEA 84.36 USD
BT222045-25G
2-Aminoacetonitrile hydrochloride ≥98%
2-Aminoacetonitrile hydrochloride

  • RTECS: MC1950000
  • Hygroscopic
  • Powder

Formula: C₂H₅ClN₂
MW: 92.53 g/mol
Melting Pt: 166…170 °C (dec.)
MDL Number: MFCD00012850
CAS Number: 6011-14-9
EINECS: 227-865-9
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR