Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Heptane-[D₁₆] ≥98% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: n-Heptane-[²H₁₆]

221895-1G
BT221895-1GEA 167.82 USD InStock
BT221895-1G
n-Heptane-[D₁₆] ≥98% (Isotopic)
n-Heptane-[D16]
CAS: 33838-52-7; EC No: 205-563-8; MDL No: MFCD00062744; RTECS: MI7700000
UN No: UN1206; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CD3(CD2)5CD3; Molecular Formula: C7D16; MW: 116.33
Melting Point: -91°; Boiling Point: 97-99°; Flash point: -4°C (24°F)
Density (g/mL): 0.794; Refractive Index: 1.384
Hygroscopic


Formula: D₃C(CD₂)₅CD₃
MW: 116,08 g/mol
Storage Temperature: ambient
MDL Number: MFCD00062744
CAS Number: 33838-52-7
EINECS: 000-000-0
UN: 1206
ADR: 3,II
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty