Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-4-methoxybenzenesulfonic acid ≥98%

Supplier: BeanTown Chemical


221800-25G
BT221800-25GEA 220.76 USD InStock
BT221800-25G
3-Amino-4-methoxybenzenesulfonic acid ≥98%
3-Amino-4-methoxybenzenesulfonic acid
CAS: 98-42-0; EC No: 202-667-5; MDL No: MFCD00035759
UN No: UN2585; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: C6H3(NH2)(OCH3)SO3H; Molecular Formula: C7H9NO4S; MW: 203.22
Melting Point: 305° (decomposes)


Formula: C₇H₉NO₄S
MDL Number: MFCD00035759
CAS Number: 98-42-0
EINECS: 202-667-5
UN: 2585
ADR: 8,III
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty