Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium on carbon 10% Type 487

Supplier: BeanTown Chemical


221570-25G 221570-5G
BT221570-25GEA 208.88 USD InStock
BT221570-25G BT221570-5G
Palladium on carbon 10% Type 487
Palladium on carbon
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Pd; MW: 106.42
Melting Point: 1555°; Boiling Point: 2963°


Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty