Pentamethylcyclopentadienyliron dicarbonyl dimer
Supplier: BeanTown Chemical
221565-1G
221565-5G
BT221565-5GEA
417.54
USD
BT221565-5G
BT221565-1G
Pentamethylcyclopentadienyliron dicarbonyl dimer
Pentamethylcyclopentadienyliron dicarbonyl dimer
- Crystalline
Formula:
C₂₄H₃₀Fe₂O₄ |
MDL Number:
MFCD00058862 CAS Number: 35344-11-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...