1,2-Epoxycyclopentane ≥97%

Supplier: BeanTown Chemical

Synonyms: Cyclopentene oxide

221475-5G 221475-25G
BT221475-5GEA 74.06 USD
BT221475-5G BT221475-25G
1,2-Epoxycyclopentane ≥97%
1,2-Epoxycyclopentane

  • RTECS: RN8935000
  • Refractive index: 1.434
  • Liquid

Formula: C₅H₈O
MW: 84.12
Boiling Pt: 102 °C
Density: 0.964 g/mL
Flash Pt: 10 °C (50 °F)
MDL Number: MFCD00005161
CAS Number: 285-67-6
EINECS: 206-005-6
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR