Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium fluoride, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical


Ampuled under argon

221415-10G 221415-50G
BT221415-10GEA 121.64 USD InStock
BT221415-10G BT221415-50G
Lithium fluoride, anhydrous ≥99.99% (trace metals basis)
Lithium fluoride
CAS: 7789-24-4; EC No: 232-152-0; MDL No: MFCD00011090; RTECS: OJ6125000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: LiF; MW: 25.94
Melting Point: 845°; Boiling Point: 1673°
Density (g/mL): 2.64


Formula: FLi
MW: 25,94 g/mol
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 870 °C
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553
Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty