Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-2-(trifluoromethyl)phenyl isocyanate ≥97%

Supplier: BeanTown Chemical


221405-1G 221405-5G
BT221405-1GEA 29.07 USD InStock
BT221405-1G BT221405-5G
4-Chloro-2-(trifluoromethyl)phenyl isocyanate ≥97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate
CAS: 16588-69-5; MDL No: MFCD00013873
UN No: UN2206; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C8H3ClF3NO; MW: 221.57
Boiling Point: 113-115°/10 mmHg; Flash point: 94°C (201°F)
Density (g/mL): 1.467; Refractive Index: 1.501
Moisture Sensitive


Formula: C₈H₃ClF₃NO
MW: 221,57 g/mol
Boiling Pt: 85 to 90 °C (13 torr)
Density: 1.4666 g/cm³ (20 °C)
MDL Number: MFCD00013873
CAS Number: 16588-69-5
EINECS: 000-000-0
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty