Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium oxide ≥99.5% (trace metals basis) -100 mesh

Supplier: BeanTown Chemical


221370-25G 221370-5G 221370-100G
BT221370-25GEA 104.23 USD InStock
BT221370-25G BT221370-5G BT221370-100G
Strontium oxide ≥99.5% (trace metals basis) -100 mesh
Strontium oxide
CAS: 1314-11-0; EC No: 215-219-9; MDL No: MFCD00049558
UN No: UN3262; Haz Class: 8; Packing Group: II
Powder; Linear Formula: SrO; MW: 103.62
Melting Point: 2430°; Boiling Point: 3000°
Density (g/mL): 4.7
Moisture Sensitive


Formula: SrO
MW: 103,62 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 2430 °C
Density: 4.7 g/cm³ (20 °C)
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 12,09009
Order Now
Specification Test Results
Assay 99.5%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty