Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethyl orthopropionate ≥95%

Supplier: BeanTown Chemical


221345-25ML
BT221345-25MLEA 139.65 USD InStock
BT221345-25ML
Trimethyl orthopropionate ≥95%
Trimethyl orthopropionate
CAS: 24823-81-2; MDL No: MFCD00043305
UN No: UN3272; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2C(OCH3)3; Molecular Formula: C6H14O3; MW: 134.17
Boiling Point: 121-122°; Flash point: 19°C (66°F)
Density (g/mL): 0.944; Refractive Index: 1.398


Formula: C₆H₁₄O₃
MW: 134,18 g/mol
MDL Number: MFCD00043305
CAS Number: 24823-81-2
EINECS: 000-000-0
UN: 3272
ADR: 3,II
Order Now
Specification Test Results
Assay 95%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty