Acetic acid glacial ≥99.7% ACS

Supplier: BeanTown Chemical

Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

221320-500ML 221320-2L 221320-100ML
BT221320-500MLEA 39.34 USD
BT221320-500ML BT221320-2L BT221320-100ML
Acetic acid glacial ≥99.7% ACS
Acetic acid

  • Refractive index: 1.371
  • Liquid

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 117…118 °C
Melting Pt: 16.2 °C
Density: 1.049 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Specification Test Results

Assay 99.7% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR