Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrocatechol ≥99%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: o-DihydroxybenzeneCatechol1,2-Dihydroxybenzene

221295-1KG 221295-5KG 221295-250G
BT221295-1KGEA 54.12 USD InStock
BT221295-1KG BT221295-5KG BT221295-250G
Pyrocatechol ≥99%, Technical Grade
Pyrocatechol
CAS: 120-80-9; EC No: 204-427-5; MDL No: MFCD00002188; RTECS: UX1050000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H6O2; MW: 110.11
Melting Point: 100-103°; Boiling Point: 245°; Flash point: 127°C (261°F)


Formula: C₆H₆O₂
MW: 110,11 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 103 to 105 °C
Density: 1.0548 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08089
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty