Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) tetrafluoroborate hexahydrate ≥98%

Supplier: BeanTown Chemical


221270-100G 221270-500G
BT221270-100GEA 73.11 USD InStock
BT221270-100G BT221270-500G
Copper(II) tetrafluoroborate hexahydrate ≥98%
Copper(II) tetrafluoroborate hexahydrate
CAS: 72259-10-0; EC No: 253-959-4; MDL No: MFCD00150223
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: Cu(BF4)2·6H2O; MW: 345.25
Hygroscopic


Formula: B₂CuF₈H₁₂O₆
MW: 345,243 g/mol
CAS Number: 72259-10-0
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty