Lanthanum, compound with nickel (1:5) ≥99.9% (by rare eath metals basis)
Supplier: BeanTown Chemical
Synonyms:
Lanthanum nickel
221200-50G
BT221200-50GEA
112.15
USD
BT221200-50G
Lanthanum, compound with nickel (1:5) ≥99.9% (by rare eath metals basis)
Lanthanum, compound with nickel (1:5)
- Pieces, Intermetallic, packaged under argon
- Air Sensitive, Moisture Sensitive
Formula:
LaNi₅ MW: 432.36 g/mol |
MDL Number:
MFCD00198177 CAS Number: 12196-72-4 EINECS: 235-372-5 UN: 3178 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...