Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-o-toluic acid ≥97%

Supplier: BeanTown Chemical


221100-10G 221100-50G
BT221100-10GEA 103.29 USD InStock
BT221100-10G BT221100-50G
α-Bromo-o-toluic acid ≥97%
α-Bromo-o-toluic acid
CAS: 7115-89-1; MDL No: MFCD00090420
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: C8H7BrO2; MW: 215.04


Formula: C₈H₇BrO₂
MW: 215,05 g/mol
MDL Number: MFCD00090420
CAS Number: 7115-89-1
EINECS: 000-000-0
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty