Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicyclopentadiene ≥95%

Supplier: BeanTown Chemical

Synonyms: 3a,4,7,7a-Tetrahydro-4,7-methanoindene

221095-250G 221095-1KG
BT221095-250GEA 18.94 USD InStock
BT221095-250G BT221095-1KG
Dicyclopentadiene ≥95%
Dicyclopentadiene
CAS: 77-73-6; EC No: 201-052-9; MDL No: MFCD00078246; RTECS: PC1050000
UN No: UN2048; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C10H12; MW: 132.21
Boiling Point: 170°; Flash point: 32°C (90°F)
Density (g/mL): 0.986


Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: 12 to 34 °C
Density: 0.982 g/cm³ (20 °C)
Flash Pt: 27 °C ()
Storage Temperature: ambient
MDL Number: MFCD00078246
CAS Number: 77-73-6
EINECS: 201-052-9
UN: 2048
ADR: 3,III
Merck Index: 13,02767
Order Now
Specification Test Results
Assay 95%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty