Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cesium ≥99.98% (trace metals basis)

Supplier: BeanTown Chemical


221025-25G 221025-1G 221025-5G
BT221025-25GEA 548.49 USD InStock
BT221025-25G BT221025-1G BT221025-5G
Cesium ≥99.98% (trace metals basis)
Cesium
CAS: 7440-46-2; EC No: 231-155-4; MDL No: MFCD00134037; RTECS: FK9225000
UN No: UN1407; Haz Class: 4.3; Packing Group: I
Solid, vacuum sealed in breakseal ampules ; Molecular Formula: Cs; MW: 132.91
Melting Point: 28.5°; Boiling Point: 705°
Density (g/mL): 1.873


Formula: Cs
MW: 132,91 g/mol
Boiling Pt: 705 °C (1013 hPa)
Melting Pt: 28.5 °C
Density: 1.873 g/cm³ (20 °C)
MDL Number: MFCD00134037
CAS Number: 7440-46-2
EINECS: 231-155-4
UN: 1407
ADR: 4.3,I
Merck Index: 12,02051
Order Now
Specification Test Results
Assay 99.98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty