Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yttrium(III) isopropoxide ≥90%, Technical Grade

Supplier: BeanTown Chemical


220905-5G 220905-1G
BT220905-5GEA 245.53 USD InStock
BT220905-5G BT220905-1G
Yttrium(III) isopropoxide ≥90%, Technical Grade
Yttrium(III) isopropoxide
CAS: 2172-12-5; MDL No: MFCD00015641
UN No: UN2925; Haz Class: 4.1 (8); Packing Group: III
Powder; Linear Formula: Y[OCH(CH3)2]3; MW: 266.17
Melting Point: 160°; Boiling Point: 200°/0.1 mmHg
Moisture Sensitive


Formula: C₉H₂₁O₃Y
MW: 266,17 gf/mol
MDL Number: MFCD00015641
CAS Number: 2172-12-5
Order Now
Specification Test Results
Assay 90%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty