Triethylammonium dihydrogentrifluoride

Supplier: BeanTown Chemical

Synonyms: Triethylamine trishydrofluorideTriethylamine trihydrofluoride

220755-5G 220755-25G 220755-100G
BT220755-5GEA 20.84 USD
BT220755-5G BT220755-100G BT220755-25G
Triethylammonium dihydrogentrifluoride
Triethylammonium dihydrogentrifluoride

  • Refractive index: 1.392
  • Hygroscopic
  • Liquid

Formula: C₆H₁₈F₃N
MW: 161.21 g/mol
Boiling Pt: 70 °C/15 mmHg
Melting Pt: –29 °C
Density: 0.989 g/mL
Flash Pt: 87 °C (188 °F)
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 1790
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR