Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indium(III) iodide, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical


220680-5G 220680-1G
BT220680-5GEA 95.89 USD InStock
BT220680-5G BT220680-1G
Indium(III) iodide, anhydrous ≥99.999% (trace metals basis)
Indium (III) iodide
CAS: 13510-35-5; EC No: 236-839-6; MDL No: MFCD00016151
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: InI3; MW: 495.53
Density (g/mL): 4.69
Hygroscopic , Light Sensitive


Formula: I₃In
MW: 495,53 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 209.85 °C
Density: 4.69 g/cm³ (20 °C)
MDL Number: MFCD00016151
CAS Number: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8,III
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty