(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene

Supplier: BeanTown Chemical

Synonyms: (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole(S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine(S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole(S)-Me-CBS Catalyst

220635-5ML 220635-1ML 220635-25ML
BT220635-1MLEA 41.45 USD
BT220635-1ML BT220635-5ML BT220635-25ML
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(S)-(-)-2-Methyl-CBS-oxazaborolidine

  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
Density: 0.929 g/mL
Flash Pt: 4 °C (39 °F)
MDL Number: MFCD00078439
CAS Number: 112022-81-8
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR