Ethyl-2-cyanoacetoacetate

Supplier: BeanTown Chemical

Synonyms: Ethyl2-acetylcyanoacetate

220590-1G 220590-5G
BT220590-1GEA 30.96 USD
BT220590-1G BT220590-5G
Ethyl-2-cyanoacetoacetate
Ethyl-2-cyanoacetoacetate

  • Liquid

Formula: C₇H₉NO₃
MW: 155.153 g/mol
MDL Number: MFCD00043487
CAS Number: 634-55-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR