Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thallium(III) chloride tetrahydrate

Supplier: BeanTown Chemical

Synonyms: Thallium trichloride tetrahydrateThallium(III) chloride hydrate

220575-25G 220575-5G
BT220575-25GEA 376.18 USD InStock
BT220575-25G BT220575-5G
Thallium(III) chloride tetrahydrate
Thallium (III) chloride tetrahydrate
CAS: 13453-33-3; EC No: 236-625-2; MDL No: MFCD00049600
UN No: UN1707; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: TlCl3·xH2O; MW: 310.73 (anhydrous)
Melting Point: 37°
Hygroscopic


Formula: Cl₃Tl·4H₂O
MW: 382,8 g/mol
MDL Number: MFCD00049600
CAS Number: 13453-33-3
EINECS: 236-625-2
UN: 1707
ADR: 6.1,II
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty