2-Heptanone ≥99%

220465-100ML 220465-500ML
BT220465-500MLEA 99.33 USD
BT220465-500ML BT220465-100ML
2-Heptanone ≥99%
2-Heptanone

  • RTECS: MJ5075000
  • Refractive index: 1.408
  • Liquid

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 149…150 °C
Melting Pt: –35 °C
Density: 0.82 g/mL
Flash Pt: 41 °C (105 °F)
MDL Number: MFCD00009513
CAS Number: 110-43-0
EINECS: 203-767-1
UN: 1110
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR