Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylsilane ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: TMS

220395-25G 220395-100G
BT220395-25GEA 41.25 USD InStock
BT220395-25G BT220395-100G
Tetramethylsilane ≥99.9% (trace metals basis)
Tetramethylsilane
CAS: 75-76-3; EC No: 200-899-1; MDL No: MFCD00008274; RTECS: VV5706000
UN No: UN2749; Haz Class: 3; Packing Group: I
Liquid; Linear Formula: Si(CH3)4; MW: 88.23
Melting Point: -99°; Boiling Point: 26-28°; Flash point: -27°C (-17°F)
Density (g/mL): 0.648; Refractive Index: 1.358


Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: -99 °C
Density: 0.642 g/cm³ (20 °C)
Flash Pt: -54 °C
Storage Temperature: 2 - 8°C
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty