Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether ≥99.0% inhibitor-free for spectrophotometry

Supplier: BeanTown Chemical

Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

219890-1L 219890-4X1L 219890-4L
BT219890-1LEA 56.04 USD InStock
BT219890-1L BT219890-4X1L BT219890-4L
Diethyl ether ≥99.0% inhibitor-free for spectrophotometry
Diethyl ether
CAS: 60-29-7; EC No: 200-467-2; MDL No: MFCD00011646; RTECS: KI5775000
UN No: UN1155; Haz Class: 3; Packing Group: I
Liquid; Linear Formula: (CH3CH2)2O; MW: 74.12
Melting Point: -116°; Boiling Point: 34.6°; Flash point: -40°C (-40°F)
Density (g/mL): 0.706; Refractive Index: 1.353


Formula: (CH₃CH₂)₂O
MW: 74,12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: -123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: -40 °C
Storage Temperature: ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Assay 99.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty