Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.7% for spectrophotometry

Supplier: BeanTown Chemical

Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

219850-1L 219850-4L 219850-4X1L
BT219850-1LEA 39.79 USD InStock
BT219850-1L BT219850-4L BT219850-4X1L
2-Propanol ≥99.7% for spectrophotometry
2-Propanol
CAS: 67-63-0; EC No: 200-661-7; MDL No: MFCD00011674; RTECS: NT8050000
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHOH; Molecular Formula: C3H8O; MW: 60.10
Melting Point: -90°; Boiling Point: 82°; Flash point: 12°C (54°F)
Density (g/mL): 0.785; Refractive Index: 1.377


Formula: (CH₃)₂CHOH
MW: 60,1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Assay 99.7% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty