Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitroguanidine

Supplier: BeanTown Chemical

Synonyms: N-Nitroguanidine

219790-100G 219790-500G
BT219790-100GEA 28.99 USD InStock
BT219790-100G BT219790-500G
Nitroguanidine
Nitroguanidine
CAS: 556-88-7; MDL No: MFCD00007039; RTECS: MF4600000
UN No: UN1336; Haz Class: 4.1; Packing Group: I
Powder; Linear Formula: NO2NHC(NH2)=NH; Molecular Formula: CH4N4O2; MW: 104.07
Melting Point: 239° (decomposes)


Formula: CH₄N₄O₂
MW: 104,07 g/mol
MDL Number: MFCD00007039
CAS Number: 556-88-7
EINECS: 209-143-5
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty