Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Trimethyl-1H-pyrazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,3,5-Trimethyl-1H-pyrazole1,3,5-Trimethylpyrazole

219760-25G 219760-5G
BT219760-25GEA 344.63 USD InStock
BT219760-25G BT219760-5G
1,3,5-Trimethyl-1H-pyrazole ≥98%
1,3,5-Trimethyl-1H-pyrazole
CAS: 1072-91-9; MDL No: MFCD00015536
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: C6H10N2; MW: 110.16
Melting Point: 38-41°; Flash point: 40°C (104°F)


Formula: C₆H₁₀N₂
MW: 110,16 g/mol
Boiling Pt: 167 to 170 °C (1013 hPa)
Melting Pt: 30 to 35 °C
Density: 0.9269 g/cm³ (40 °C)
MDL Number: MFCD00015536
CAS Number: 1072-91-9
EINECS: 000-000-0
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty