Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium chloride, anhydrous ≥99.98% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Aluminum chloride

219670-50G 219670-250G
BT219670-50GEA 62.45 USD InStock
BT219670-50G BT219670-250G
Aluminium chloride, anhydrous ≥99.98% (trace metals basis)
Aluminium chloride
CAS: 7446-70-0; EC No: 231-208-1; MDL No: MFCD00003422; RTECS: BD0525000
UN No: UN1726; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: AlCl3; MW: 133.34
Melting Point: 190°
Moisture Sensitive


Formula: AlCl₃
MW: 133,34 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 180 to 181 °C
Density: liquid 1.31 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Merck Index: 13,00335
Order Now
Specification Test Results
Assay 99.98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty