Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform-[D₁] ≥100% (Isotopic) contains 0.03% TMS

Supplier: BeanTown Chemical

Synonyms: Chloroform-DTrichloromethane-D1Chloroform-[²H₁]

219655-10EACH 219655-5EACH
BT219655-10EAEA 74.23 USD InStock
BT219655-10EA BT219655-5EACH
Chloroform-[D₁] ≥100% (Isotopic) contains 0.03% TMS
Chloroform-[D1]
CAS: 865-49-6; EC No: 212-742-4; MDL No: MFCD00000827
UN No: UN1888; Haz Class: 6.1; Packing Group: III
Liquid, in prescored ampules, 0.75ml/ampule ; Molecular Formula: CDCl3; MW: 120.38
Melting Point: -64°; Boiling Point: 60.9°
Density (g/mL): 1.5; Refractive Index: 1.444
Light Sensitive, Moisture Sensitive


Formula: CDCl₃
MW: 120,38 g/mol
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: -64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: 2 - 8°C
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III
Order Now
Specification Test Results
Assay 100%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty