Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yttrium(III) fluoride, anhydrous ≥99.99% (trace rare earth metals basis)

Supplier: BeanTown Chemical


219645-100G 219645-500G 219645-25G
BT219645-100GEA 305.22 USD InStock
BT219645-100G BT219645-500G BT219645-25G
Yttrium(III) fluoride, anhydrous ≥99.99% (trace rare earth metals basis)
Yttrium (III) fluoride
CAS: 13709-49-4; EC No: 237-257-5; MDL No: MFCD00049714
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder/Lump; Linear Formula: YF3; MW: 145.9
Melting Point: 1387°
Density (g/mL): 4.01
Hygroscopic


Formula: F₃Y
MW: 145,9 g/mol
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty