Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dirhenium heptaoxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical


Ampuled under argon

219620-5G 219620-1G
BT219620-5GEA 157.67 USD InStock
BT219620-5G BT219620-1G
Dirhenium heptaoxide ≥99.9% (trace metals basis)
di-Rhenium heptaoxide
CAS: 1314-68-7; EC No: 215-241-9; MDL No: MFCD00011199
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III
Crystalline; Linear Formula: Re2O7; MW: 484.4
Melting Point: 220°; Boiling Point: 360°
Density (g/mL): 6.103
Moisture Sensitive


Formula: O₇Re₂
MW: 484,41 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 220 °C
Density: 6 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011199
CAS Number: 1314-68-7
EINECS: 215-241-9
UN: 3085
ADR: 5.1,II
Merck Index: 12,08344
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty