Tris(dimethylamino)phosphine ≥97%
Supplier: BeanTown Chemical
Synonyms:
Hexamethyl phosphorous triamide
, Tris(dimethylamino)phosphine
, HMPT
, Hexamethyltriaminophosphine
219550-5G
219550-25G
BT219550-5GEA
58.04
USD
BT219550-5G
BT219550-25G
Tris(dimethylamino)phosphine ≥97%
Tris(dimethylamino)phosphine
- RTECS: TH3390000
- Refractive index: 1.463
- Air Sensitive
- Liquid
Formula:
C₆H₁₈N₃P MW: 163.2 g/mol Boiling Pt: 48…50 °C/12 mmHg Density: 0.898 g/mL Flash Pt: 27 °C (81 °F) |
MDL Number:
MFCD00008301 CAS Number: 1608-26-0 EINECS: 216-534-4 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...