1,3-Dichloropropane ≥95%

Supplier: BeanTown Chemical

219510-25G 219510-100G
BT219510-100GEA 316.39 USD
BT219510-100G BT219510-25G
1,3-Dichloropropane ≥95%
1,3-Dichloropropane

  • Refractive index: 1.448
  • Liquid

Formula: ClH₂CCH₂CH₂Cl
MW: 112.99 g/mol
Boiling Pt: 120…122 °C
Melting Pt: –99 °C
Density: 1.19 g/mL
Flash Pt: 36 °C (97 °F)
MDL Number: MFCD00000999
CAS Number: 142-28-9
EINECS: 205-531-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR