Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum(III) 2-ethylhexanoate 10% (w/v) in hexane

Supplier: BeanTown Chemical


219360-25ML 219360-100ML
BT219360-25MLEA 175.07 USD InStock
BT219360-25ML BT219360-100ML
Lanthanum(III) 2-ethylhexanoate 10% (w/v) in hexane
Lanthanum(III) 2-ethylhexanoate
CAS: 67816-09-5; EC No: 267-212-5; MDL No: MFCD00798555
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C24H45LaO6 ; MW: 568.18


Formula: C₂₄H₄₅LaO₆
MW: 568,523 g/mol
MDL Number: MFCD00798555
CAS Number: 67816-09-5
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty