Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroacetic acid ≥99%

Supplier: BeanTown Chemical

Danger
Synonyms: TCAAceto-CaustinTrichloroethanoic acid

219325-1KG 219325-250G
BT219325-1KGEA 88.64 USD InStock
BT219325-1KG BT219325-250G
Trichloroacetic acid ≥99%
Trichloroacetic acid (glacial)
CAS: 76-03-9; EC No: 200-927-2; MDL No: MFCD00004177; RTECS: AJ7875000
UN No: UN1839; Haz Class: 8; Packing Group: II
Crystalline/Powder; Linear Formula: CCl3COOH; MW: 163.39
Melting Point: 54-58°; Boiling Point: 196°; Flash point: 110°C (230°F)
Density (g/mL): 1.629; Refractive Index: 1.62
Hygroscopic


Formula: Cl₃CCOOH
MW: 163,39 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54 to 56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 110 °C (closed cup)
Storage Temperature: ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://us.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty