2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95%

Synonyms: Ethylene glycol chlorophosphate2-Chloro-1,3,2-dioxaphospholane-2-oxide

219190-100G 219190-25G 219190-5G
BT219190-100GEA 796.64 USD
BT219190-100G BT219190-25G BT219190-5G
2-Chloro-2-oxo-1,3,2-dioxaphospholane ≥95%
2-Chloro-2-oxo-1,3,2-dioxaphospholane

  • Refractive index: 1.450
  • Moisture Sensitive
  • Liquid

Formula: C₂H₄ClO₃P
MW: 142.47 g/mol
Boiling Pt: 89…91 °C/0.8 mmHg
Melting Pt: 12…14 °C
Density: 1.55 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00043138
CAS Number: 6609-64-9
EINECS: 229-560-6
ADR: 8,II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR