Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium nitrate, anhydrous ≥99.97% (trace metals basis)

Supplier: BeanTown Chemical


219175-100G 219175-25G
BT219175-100GEA 305.22 USD InStock
BT219175-100G BT219175-25G
Strontium nitrate, anhydrous ≥99.97% (trace metals basis)
Strontium nitrate
CAS: 10042-76-9; EC No: 233-131-9; MDL No: MFCD00011248; RTECS: WK9800000
UN No: UN1507; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Sr(NO3)2; MW: 211.63
Melting Point: 570°
Density (g/mL): 2.99
Hygroscopic


Formula: Sr(NO₃)₂
MW: 211,63 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 13,08926
Order Now
Specification Test Results
Assay 99.97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty