Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt ≥99.5% (trace metals basis), Powder -325 mesh

Supplier: BeanTown Chemical


219080-100G 219080-500G
BT219080-100GEA 94.8 USD InStock
BT219080-100G BT219080-500G
Cobalt ≥99.5% (trace metals basis), Powder -325 mesh
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92


Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II
Merck Index: 12,02488
Order Now
Specification Test Results
Assay 99.5%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty