Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethyl orthosilicate (TEOS) ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Tetraethoxysilane

218985-2KG 218985-500G 218985-100G
BT218985-2KGEA 176.4 USD InStock
BT218985-2KG BT218985-500G BT218985-100G
Tetraethyl orthosilicate (TEOS) ≥99.9% (trace metals basis)
Tetraethyl orthosilicate (TEOS)
CAS: 78-10-4; EC No: 201-083-8; MDL No: MFCD00009062; RTECS: VV9450000
UN No: UN1292; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: Si(OCH2CH3)4; Molecular Formula: C8H20O4Si; MW: 208.33
Melting Point: -85°; Boiling Point: 168°; Flash point: 46°C (116°F)
Density (g/mL): 0.934; Refractive Index: 1.382
Moisture Sensitive


Formula: Si(OC₂H₅)₄
MW: 208,33 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: -85.5 °C
Density: 0.933 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty