Pyridine ≥99.5% for spectrophotometry

218920-250ML 218920-4L 218920-1L
BT218920-250MLEA 48.7 USD
BT218920-250ML BT218920-1L BT218920-4L
Pyridine ≥99.5% for spectrophotometry
Pyridine

  • RTECS: UR8400000
  • Refractive index: 1.509
  • Liquid

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C
Melting Pt: –42 °C
Density: 0.978 g/mL
Flash Pt: 17 °C (62.6 °F)
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg

Order Now

Specification Test Results

Assay 99.5% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR