BEAN218685

218685-25G 218685-1G 218685-5G
BT218685-1GEA 63.14 USD
BT218685-1G BT218685-5G BT218685-25G
BEAN218685
  • RTECS: RQ7350000
  • Refractive index: 1.412
  • Moisture Sensitive
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR