Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid 2.0 N in water

Supplier: BeanTown Chemical

Danger
Synonyms: Methanecarboxylic acidGlacial acetic acidVinegar acidEthanoic acid

218570-20L 218570-4L 218570-1L
BT218570-20LEA 123.89 USD InStock
BT218570-20L BT218570-4L BT218570-1L
Acetic acid 2.0 N in water
Acetic acid
CAS: 64-19-7; EC No: 200-580-7; MDL No: MFCD00036152
UN No: UN2790; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: CH3CO2H ; MW: 60.05


Formula: H₃CCOOH
MW: 60,05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: 15 - 25°C
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
REACH: 01-2119475328-30
https://us.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty