Sulfuryl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Sulphuryl dichlorideSulfuryl dichloride

218545-250G 218545-1KG
BT218545-250GEA 212.15 USD
BT218545-250G BT218545-1KG
Sulfuryl chloride ≥97%
Sulphuryl chloride

  • RTECS: WT4870000
  • Refractive index: 1.443
  • Moisture Sensitive
  • Liquid

Formula: Cl₂O₂S
MW: 134.97 g/mol
Boiling Pt: 68…70 °C
Melting Pt: –54 °C
Density: 1.665 g/mL
MDL Number: MFCD00011451
CAS Number: 7791-25-5
EINECS: 232-245-6
UN: 1834
ADR: 8,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR