Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) nitrate hydrate ≥99.99% (trace rare earth metals basis)

Supplier: BeanTown Chemical


218510-10G 218510-50G
BT218510-10GEA 84.14 USD InStock
BT218510-10G BT218510-50G
Ytterbium(III) nitrate hydrate ≥99.99% (trace rare earth metals basis)
Ytterbium(III) nitrate hydrate
EC No: 237-384-6; MDL No: MFCD00149883
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: Yb(NO3)3·xH2O; MW: 359.05 (anhydrous)
Hygroscopic


Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty