Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium hexacarbonyl ≥99%

Supplier: BeanTown Chemical

Synonyms: Chromium carbonylHexacarbonylchromiumChromium(0) hexacarbonylHexacarbonylchromium(0)

218475-10G 218475-50G
BT218475-10GEA 86.37 USD InStock
BT218475-10G BT218475-50G
Chromium hexacarbonyl ≥99%
Chromium hexacarbonyl
CAS: 13007-92-6; EC No: 235-852-4; MDL No: MFCD00010945; RTECS: GB5075000
UN No: UN3466; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: Cr(CO)6; MW: 220.06
Melting Point: 150° (decomposes)
Density (g/mL): 1.77
Light Sensitive


Formula: C₆CrO₆
MW: 220,06 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 90 °C
Density: 1.77 g/cm³ (20 °C)
MDL Number: MFCD00010945
CAS Number: 13007-92-6
EINECS: 235-852-4
UN: 3466
ADR: 6.1,II
Merck Index: 13,02253
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty