Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tantalum(V) ethanolate ≥99%

Supplier: BeanTown Chemical

Synonyms: Tantalum(V) ethoxide

218375-25G 218375-5G
BT218375-25GEA 317.61 USD InStock
BT218375-25G BT218375-5G
Tantalum(V) ethanolate ≥99%
Tantalum(V) ethanolate
CAS: 6074-84-6; EC No: 228-010-2; MDL No: MFCD00049785
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: (CH3CH2O)5Ta; Molecular Formula: C10H25O5Ta; MW: 406.25
Melting Point: 20°; Boiling Point: 145°/0.1 mmHg; Flash point: 29°C (84°F)
Density (g/mL): 1.566; Refractive Index: 1.487
Air Sensitive, Moisture Sensitive


Formula: C₁₀H₂₅O₅Ta
MW: 406,25 g/mol
MDL Number: MFCD00049785
CAS Number: 6074-84-6
EINECS: 228-010-2
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty