Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanesulfonyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Mesyl chloride

218300-100G 218300-500G
BT218300-100GEA 15.58 USD InStock
BT218300-100G BT218300-500G
Methanesulfonyl chloride ≥98%
Methanesulphonyl chloride
CAS: 124-63-0; EC No: 204-706-1; MDL No: MFCD00007454; RTECS: PB2790000
UN No: UN3246; Haz Class: 6.1 (8); Packing Group: I
Liquid; Linear Formula: CH3SO2Cl; MW: 114.55
Melting Point: -33°; Boiling Point: 161°; Flash point: 97°C (206°F)
Density (g/mL): 1.477; Refractive Index: 1.452
Moisture Sensitive


Formula: CH₃ClO₂S
MW: 114,55 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: -32 °C
Density: 1.478 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: ambient
MDL Number: MFCD00007454
CAS Number: 124-63-0
EINECS: 204-706-1
UN: 3246
ADR: 6.1,I
Merck Index: 13,05982
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty